accessibility menu, dialog, popup

UserName

CAS RN 3698-89-3

IUPAC Name: 1-(methylsulfanyl)dodecane
Synonyms: 1-(methylsulfanyl)dodecane
Molecular Weight (g/mol): 216.43
Molecular Formula: C13H28S
InChi Key: KJWHJDGMOQJLGF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCSC
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)