missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ n-Dodecyl-β-D-maltoside, ULTROL™, ≥99%, Calbiochem™,
$948.00 - $9,520.00
Chemical Identifiers
| CAS | 69227-93-6 |
|---|---|
| Molecular Formula | C24H46O11 |
| Molecular Weight (g/mol) | 510.621 |
| InChI Key | NLEBIOOXCVAHBD-QKMCSOCLSA-N |
| PubChem CID | 114880 |
| ChEBI | CHEBI:43769 |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
| SMILES | CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
3243555GM
|
MilliporeSigma™
3243555GM |
5 g | Glass Bottle |
Each for $948.00
|
|
||||
|
32435525GM
|
MilliporeSigma™
32435525GM |
25 g | Glass Bottle |
Each for $3,600.00
|
|
||||
|
324355100GM
|
MilliporeSigma™
324355100GM |
100 g | Glass Bottle |
Each for $5,100.00
|
|
||||
|
324355250GM
|
MilliporeSigma™
324355250GM |
250 g | Glass Bottle |
Each for $9,520.00
|
|
||||
Chemical Identifiers
| 69227-93-6 | |
| 510.621 | |
| 114880 | |
| (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
| C24H46O11 | |
| NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
| CHEBI:43769 | |
| CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O |
Specifications
| White | |
| C24H46O11 | |
| CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O | |
| 510.621 | |
| CHEBI:43769 |
| 69227-93-6 | |
| NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
| (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 114880 | |
| ≥98% (HPLC) |