missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ n-Dodecyl-β-D-maltoside, ULTROL™, ≥99%, Calbiochem™,
Supplier: MilliporeSigma™ 324355100GM
| Quantity | 100 g |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| 69227-93-6 | |
| 510.621 | |
| 114880 | |
| (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
| C24H46O11 | |
| NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
| CHEBI:43769 | |
| CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O |
Specifications
| White | |
| 100 g | |
| C24H46O11 | |
| CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O | |
| 510.621 | |
| CHEBI:43769 |
| 69227-93-6 | |
| Glass Bottle | |
| NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
| (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 114880 | |
| ≥98% (HPLC) |