missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl acrylate, tech. 90%, stab. with 60-100ppm 4-methoxyphenol

CAS: 2156-97-0 | C15H28O2 | 240.39 g/mol

Chemical Identifiers

CAS 2156-97-0
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.39
MDL Number MFCD00042871
InChI Key PBOSTUDLECTMNL-UHFFFAOYSA-N
Synonym dodecyl acrylate, lauryl acrylate, n-lauryl acrylate, n-dodecyl acrylate, 2-propenoic acid, dodecyl ester, acrylic acid, dodecyl ester, acrylic acid lauryl ester, acrylic acid dodecyl ester, unii-90ik36wgvr, 90ik36wgvr
PubChem CID 75084
IUPAC Name dodecyl prop-2-enoate
SMILES CCCCCCCCCCCCOC(=O)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH60625AE
View Documents Promotion Details
N/A 100 mL N/A
Only null left
 
AAH60625AP
View Documents Promotion Details
N/A 500 mL N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2156-97-0
240.39
PBOSTUDLECTMNL-UHFFFAOYSA-N
75084
CCCCCCCCCCCCOC(=O)C=C
C15H28O2
MFCD00042871
dodecyl acrylate, lauryl acrylate, n-lauryl acrylate, n-dodecyl acrylate, 2-propenoic acid, dodecyl ester, acrylic acid, dodecyl ester, acrylic acid lauryl ester, acrylic acid dodecyl ester, unii-90ik36wgvr, 90ik36wgvr
dodecyl prop-2-enoate

Specifications

2156-97-0
120°C (0.75 mmHg)
C15H28O2
CH2=CHCOOC12H25
UN3082
dodecyl acrylate, lauryl acrylate, n-lauryl acrylate, n-dodecyl acrylate, 2-propenoic acid, dodecyl ester, acrylic acid, dodecyl ester, acrylic acid lauryl ester, acrylic acid dodecyl ester, unii-90ik36wgvr, 90ik36wgvr
CCCCCCCCCCCCOC(=O)C=C
240.39
240.38
Technical
0.884
110°C (230°F)
1.445
MFCD00042871
1779069
PBOSTUDLECTMNL-UHFFFAOYSA-N
dodecyl prop-2-enoate
75084
90%
n-Dodecyl acrylate, stabilized with 60 to 100ppm 4-methoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 218-463-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only