accessibility menu, dialog, popup

UserName

H3COOCH2

CAS RN 2156-97-0

IUPAC Name: dodecyl prop-2-enoate
Synonyms: lauryl acrylate
Molecular Weight (g/mol): 240.39
Molecular Formula: C15H28O2
InChi Key: PBOSTUDLECTMNL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC(=O)C=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)