missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl acrylate, tech. 90%, stab. with 60-100ppm 4-methoxyphenol

Catalog No. AAH60625AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60625AE 100 mL
AAH60625AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60625AE Supplier Thermo Scientific Chemicals Supplier No. H60625AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2156-97-0 | C15H28O2 | 240.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2156-97-0
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.39
MDL Number MFCD00042871
InChI Key PBOSTUDLECTMNL-UHFFFAOYSA-N
Synonym dodecyl acrylate, lauryl acrylate, n-lauryl acrylate, n-dodecyl acrylate, 2-propenoic acid, dodecyl ester, acrylic acid, dodecyl ester, acrylic acid lauryl ester, acrylic acid dodecyl ester, unii-90ik36wgvr, 90ik36wgvr
PubChem CID 75084
IUPAC Name dodecyl prop-2-enoate
SMILES CCCCCCCCCCCCOC(=O)C=C

Specifications

Density 0.884
Boiling Point 120°C (0.75 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.445
Linear Formula CH2=CHCOOC12H25
Quantity 100 mL
UN Number UN3082
Beilstein 1779069
Formula Weight 240.38
Percent Purity 90%
Grade Technical
Chemical Name or Material n-Dodecyl acrylate, stabilized with 60 to 100ppm 4-methoxyphenol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 218-463-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only