missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl acrylate, tech. 90%, stab. with 60-100ppm 4-methoxyphenol

CAS: 2156-97-0 | C15H28O2 | 240.39 g/mol

Supplier:  Thermo Scientific Chemicals H60625AP

 View more versions of this product

Catalog No. AAH60625AP


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Dodecyl acrylate
2156-97-0
120°C (0.75 mmHg)
C15H28O2
CH2=CHCOOC12H25
500 mL
1779069
PBOSTUDLECTMNL-UHFFFAOYSA-N
dodecyl prop-2-enoate
75084
90%
stabilized with 60 to 100ppm 4-methoxyphenol
0.884
110°C (230°F)
1.445
MFCD00042871
UN3082
dodecyl acrylate, lauryl acrylate, n-lauryl acrylate, n-dodecyl acrylate, 2-propenoic acid, dodecyl ester, acrylic acid, dodecyl ester, acrylic acid lauryl ester, acrylic acid dodecyl ester, unii-90ik36wgvr, 90ik36wgvr
CCCCCCCCCCCCOC(=O)C=C
240.39
240.38
Technical

Chemical Identifiers

2156-97-0
240.39
PBOSTUDLECTMNL-UHFFFAOYSA-N
75084
CCCCCCCCCCCCOC(=O)C=C
C15H28O2
MFCD00042871
dodecyl acrylate, lauryl acrylate, n-lauryl acrylate, n-dodecyl acrylate, 2-propenoic acid, dodecyl ester, acrylic acid, dodecyl ester, acrylic acid lauryl ester, acrylic acid dodecyl ester, unii-90ik36wgvr, 90ik36wgvr
dodecyl prop-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 218-463-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only