missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Diphenylmethylene)glycine methyl ester, 98%
CAS: 81167-39-7 | C16H15NO2 | 253.301 g/mol
$124.81 - $390.14
Chemical Identifiers
| CAS | 81167-39-7 |
|---|---|
| Molecular Formula | C16H15NO2 |
| Molecular Weight (g/mol) | 253.301 |
| MDL Number | MFCD08272233 |
| InChI Key | PQTOLHHWLUCKSB-UHFFFAOYSA-N |
| Synonym | n-diphenylmethylene glycine methyl ester, methyl n-diphenylmethylene glycinate, methyl 2-diphenylmethylene amino acetate, diphenylmethylene-glycine methyl ester, methyl 2-benzhydrylideneamino acetate, methyl diphenylmethylene amino acetate, methyl 2-diphenylmethylidene amino acetate, n-diphenylmethylideneglycine methyl ester, n,n-diphenylmethylgene glycine methyl ester, benzhydrylideneamino acetic acid methyl ester |
| PubChem CID | 563248 |
| IUPAC Name | methyl 2-(benzhydrylideneamino)acetate |
| SMILES | COC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6661414
|
Thermo Scientific Chemicals
H6661414 |
25 g |
N/A
|
|
|||||
|
AAH6661422
|
Thermo Scientific Chemicals
H6661422 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 81167-39-7 | |
| 253.301 | |
| PQTOLHHWLUCKSB-UHFFFAOYSA-N | |
| 563248 | |
| COC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2 |
| C16H15NO2 | |
| MFCD08272233 | |
| n-diphenylmethylene glycine methyl ester, methyl n-diphenylmethylene glycinate, methyl 2-diphenylmethylene amino acetate, diphenylmethylene-glycine methyl ester, methyl 2-benzhydrylideneamino acetate, methyl diphenylmethylene amino acetate, methyl 2-diphenylmethylidene amino acetate, n-diphenylmethylideneglycine methyl ester, n,n-diphenylmethylgene glycine methyl ester, benzhydrylideneamino acetic acid methyl ester | |
| methyl 2-(benzhydrylideneamino)acetate |
Specifications
| 42°C to 44°C | |
| White to Yellow | |
| MFCD08272233 | |
| n-diphenylmethylene glycine methyl ester, methyl n-diphenylmethylene glycinate, methyl 2-diphenylmethylene amino acetate, diphenylmethylene-glycine methyl ester, methyl 2-benzhydrylideneamino acetate, methyl diphenylmethylene amino acetate, methyl 2-diphenylmethylidene amino acetate, n-diphenylmethylideneglycine methyl ester, n,n-diphenylmethylgene glycine methyl ester, benzhydrylideneamino acetic acid methyl ester | |
| COC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2 | |
| 25 g | |
| 563248 | |
| 98% | |
| N-(Diphenylmethylene)glycine methyl ester |
| 81167-39-7 | |
| C16H15NO2 | |
| 9695211 | |
| PQTOLHHWLUCKSB-UHFFFAOYSA-N | |
| methyl 2-(benzhydrylideneamino)acetate | |
| 253.301 | |
| 253.3 | |
| Solid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only