missing translation for 'onlineSavingsMsg'
Learn More

N-(Diphenylmethylene)glycine methyl ester, 98%

CAS: 81167-39-7 | C16H15NO2 | 253.301 g/mol

Supplier:  Thermo Scientific Chemicals H6661422

 View more versions of this product

Catalog No. AAH6661422


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

42°C to 44°C
81167-39-7
C16H15NO2
9695211
PQTOLHHWLUCKSB-UHFFFAOYSA-N
methyl 2-(benzhydrylideneamino)acetate
253.301
253.3
Solid
N-(Diphenylmethylene)glycine methyl ester
White to Yellow
MFCD08272233
n-diphenylmethylene glycine methyl ester, methyl n-diphenylmethylene glycinate, methyl 2-diphenylmethylene amino acetate, diphenylmethylene-glycine methyl ester, methyl 2-benzhydrylideneamino acetate, methyl diphenylmethylene amino acetate, methyl 2-diphenylmethylidene amino acetate, n-diphenylmethylideneglycine methyl ester, n,n-diphenylmethylgene glycine methyl ester, benzhydrylideneamino acetic acid methyl ester
COC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2
100 g
563248
98%

Chemical Identifiers

81167-39-7
253.301
PQTOLHHWLUCKSB-UHFFFAOYSA-N
563248
COC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2
C16H15NO2
MFCD08272233
n-diphenylmethylene glycine methyl ester, methyl n-diphenylmethylene glycinate, methyl 2-diphenylmethylene amino acetate, diphenylmethylene-glycine methyl ester, methyl 2-benzhydrylideneamino acetate, methyl diphenylmethylene amino acetate, methyl 2-diphenylmethylidene amino acetate, n-diphenylmethylideneglycine methyl ester, n,n-diphenylmethylgene glycine methyl ester, benzhydrylideneamino acetic acid methyl ester
methyl 2-(benzhydrylideneamino)acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only