missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ n-Decyl-β-D-maltopyranoside, ULTROL™ Grade, Calbiochem™,
Chemical Identifiers
| CAS | 82494-09-5 |
|---|---|
| Molecular Formula | C22H42O11 |
| Molecular Weight (g/mol) | 482.567 |
| InChI Key | WOQQAWHSKSSAGF-WXFJLFHKSA-N |
| PubChem CID | 5288728 |
| ChEBI | CHEBI:67097 |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-decoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
| SMILES | CCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O |
Chemical Identifiers
| 82494-09-5 | |
| 482.567 | |
| 5288728 | |
| (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-decoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
| C22H42O11 | |
| WOQQAWHSKSSAGF-WXFJLFHKSA-N | |
| CHEBI:67097 | |
| CCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O |
Specifications
| 82494-09-5 | |
| WOQQAWHSKSSAGF-WXFJLFHKSA-N | |
| (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-decoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 5288728 | |
| ≥99% (TLC) |
| C22H42O11 | |
| CCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O | |
| 482.567 | |
| CHEBI:67097 |