missing translation for 'onlineSavingsMsg'
Learn More

n-Decyl acetate, 98%

CAS: 112-17-4 | C12H24O2 | 200.32 g/mol

Chemical Identifiers

CAS 112-17-4
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00026538
InChI Key NUPSHWCALHZGOV-UHFFFAOYSA-N
Synonym n-decyl acetate, acetic acid, decyl ester, n-decyl ethanoate, 1-decanol acetate, decyl ethanoate, acetate c 10, decanyl acetate, 1-decyl acetate, acetic acid n-decyl ester, decanol acetate
PubChem CID 8167
IUPAC Name decyl acetate
SMILES CCCCCCCCCCOC(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1881430
View Documents Promotion Details
N/A 250 g N/A
Only null left
 
AAA188140B
View Documents Promotion Details
N/A 1000 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-17-4
200.32
NUPSHWCALHZGOV-UHFFFAOYSA-N
8167
CCCCCCCCCCOC(C)=O
C12H24O2
MFCD00026538
n-decyl acetate, acetic acid, decyl ester, n-decyl ethanoate, 1-decanol acetate, decyl ethanoate, acetate c 10, decanyl acetate, 1-decyl acetate, acetic acid n-decyl ester, decanol acetate
decyl acetate

Specifications

112-17-4
244°C
C12H24O2
MFCD00026538
n-decyl acetate, acetic acid, decyl ester, n-decyl ethanoate, 1-decanol acetate, decyl ethanoate, acetate c 10, decanyl acetate, 1-decyl acetate, acetic acid n-decyl ester, decanol acetate
CCCCCCCCCCOC(C)=O
200.32
200.32
n-Decyl acetate
0.867
125°C (257°F)
1.427
1762123
NUPSHWCALHZGOV-UHFFFAOYSA-N
decyl acetate
8167
98%
Videos
Safety and Handling

Safety and Handling

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 203-942-2

RTECSNumber : AG5235000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only