missing translation for 'onlineSavingsMsg'
Learn More

n-Decyl acetate, 98%

CAS: 112-17-4 | C12H24O2 | 200.32 g/mol

Supplier:  Thermo Scientific Chemicals A188140B

Quantity 1000 g

 View more versions of this product

Catalog No. AAA188140B

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-17-4
200.32
NUPSHWCALHZGOV-UHFFFAOYSA-N
8167
CCCCCCCCCCOC(C)=O
C12H24O2
MFCD00026538
n-decyl acetate, acetic acid, decyl ester, n-decyl ethanoate, 1-decanol acetate, decyl ethanoate, acetate c 10, decanyl acetate, 1-decyl acetate, acetic acid n-decyl ester, decanol acetate
decyl acetate

Specifications

n-Decyl acetate
0.867
125°C (257°F)
1.427
MFCD00026538
n-decyl acetate, acetic acid, decyl ester, n-decyl ethanoate, 1-decanol acetate, decyl ethanoate, acetate c 10, decanyl acetate, 1-decyl acetate, acetic acid n-decyl ester, decanol acetate
CCCCCCCCCCOC(C)=O
200.32
200.32
112-17-4
244°C
C12H24O2
1000 g
1762123
NUPSHWCALHZGOV-UHFFFAOYSA-N
decyl acetate
8167
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 203-942-2

RTECSNumber : AG5235000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions