accessibility menu, dialog, popup

UserName

CAS RN 112-17-4

IUPAC Name: decyl acetate
Synonyms: decyl acetate
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: NUPSHWCALHZGOV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOC(C)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)