missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Cyclohexylthio)phthalimide 98.0+%, TCI America™
$72.70 - $296.38
Chemical Identifiers
| CAS | 17796-82-6 |
|---|---|
| Molecular Formula | C14H15NO2S |
| Molecular Weight (g/mol) | 261.339 |
| MDL Number | MFCD00072247 |
| InChI Key | UEZWYKZHXASYJN-UHFFFAOYSA-N |
| Synonym | Cyclohexyl N-Phthalimidyl Sulfide |
| PubChem CID | 28777 |
| IUPAC Name | 2-cyclohexylsulfanylisoindole-1,3-dione |
| SMILES | C1CCC(CC1)SN2C(=O)C3=CC=CC=C3C2=O |
Chemical Identifiers
| 17796-82-6 | |
| 261.339 | |
| UEZWYKZHXASYJN-UHFFFAOYSA-N | |
| 28777 | |
| C1CCC(CC1)SN2C(=O)C3=CC=CC=C3C2=O |
| C14H15NO2S | |
| MFCD00072247 | |
| Cyclohexyl N-Phthalimidyl Sulfide | |
| 2-cyclohexylsulfanylisoindole-1,3-dione |
Specifications
| 17796-82-6 | |
| White-Yellow | |
| MFCD00072247 | |
| Cyclohexyl N-Phthalimidyl Sulfide | |
| C1CCC(CC1)SN2C(=O)C3=CC=CC=C3C2=O | |
| 261.339 | |
| 261.34 | |
| Crystalline Powder |
| 91°C | |
| C14H15NO2S | |
| 25 g | |
| UEZWYKZHXASYJN-UHFFFAOYSA-N | |
| 2-cyclohexylsulfanylisoindole-1,3-dione | |
| 28777 | |
| ≥98.0% (N) | |
| N-(Cyclohexylthio)phthalimide |
Safety and Handling
EINECSNumber : (5)-3358
RTECSNumber : TI4290000
TSCA : Yes