missing translation for 'onlineSavingsMsg'
Learn More

N-Cyano-N',S-dimethylisothiourea 98.0+%, TCI America™

Catalog No. C112225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C112225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C112225G Supplier TCI America Supplier No. C112225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5848-24-8
Molecular Formula C4H7N3S
Molecular Weight (g/mol) 129.181
MDL Number MFCD00078154
InChI Key MHGGQXIPBPGZFB-UHFFFAOYSA-N
PubChem CID 111103
IUPAC Name methyl N-cyano-N'-methylcarbamimidothioate
SMILES CN=C(NC#N)SC

Specifications

Melting Point 203°C
Color White
Quantity 25 g
Formula Weight 129.18
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Cyano-N′,S-dimethylisothiourea