accessibility menu, dialog, popup

UserName

CAS RN 5848-24-8

IUPAC Name: N-cyano-N'-methyl(methylsulfanyl)methanimidamide
Synonyms: N-cyano-N'-methylmethylsulfanylmethanimidamide
Molecular Weight (g/mol): 129.18
Molecular Formula: C4H7N3S
InChi Key: MHGGQXIPBPGZFB-UHFFFAOYSA-N
SMILES: CSC(NC#N)=NC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results