missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-serine 98.0+%, TCI America™

$51.98 - $165.56

Chemical Identifiers

CAS 1145-80-8
Molecular Formula C11H13NO5
Molecular Weight (g/mol) 239.23
MDL Number MFCD00002662
InChI Key GNIDSOFZAKMQAO-UHFFFAOYNA-N
Synonym z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine
PubChem CID 100310
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoic acid
SMILES OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C06355G
View Documents
TCI America
C06355G
5 g
Each for $51.98
Only null left
 
C063525G
View Documents
TCI America
C063525G
25 g
Each for $165.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1145-80-8
239.23
GNIDSOFZAKMQAO-UHFFFAOYNA-N
100310
OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
C11H13NO5
MFCD00002662
z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine
2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoic acid

Specifications

1145-80-8
White
MFCD00002662
z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine
OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
239.23
239.23
Crystalline Powder
118°C
C11H13NO5
5 g
GNIDSOFZAKMQAO-UHFFFAOYNA-N
2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoic acid
100310
≥98.0% (HPLC,T)
N-Carbobenzoxy-L-serine
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents