missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-L-serine 98.0+%, TCI America™
Supplier: TCI America C063525G
Specifications
| N-Carbobenzoxy-L-serine | |
| 118°C | |
| C11H13NO5 | |
| 25 g | |
| GNIDSOFZAKMQAO-UHFFFAOYNA-N | |
| 2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoic acid | |
| 100310 | |
| ≥98.0% (HPLC,T) |
| 1145-80-8 | |
| White | |
| MFCD00002662 | |
| z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine | |
| OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O | |
| 239.23 | |
| 239.23 | |
| Crystalline Powder |
Chemical Identifiers
| 1145-80-8 | |
| 239.23 | |
| GNIDSOFZAKMQAO-UHFFFAOYNA-N | |
| 100310 | |
| OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O |
| C11H13NO5 | |
| MFCD00002662 | |
| z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine | |
| 2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoic acid |
Safety and Handling
TSCA : No