accessibility menu, dialog, popup

UserName

OHHNOOOHO

CAS RN 1145-80-8

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoic acid
Molecular Weight (g/mol): 239.23
Molecular Formula: C11H13NO5
InChi Key: GNIDSOFZAKMQAO-UHFFFAOYNA-N
SMILES: OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)