missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-leucine 96.0+%, TCI America™

Supplier:  TCI America C073925G

 View more versions of this product

Catalog No. C073925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Carbobenzoxy-L-leucine
Yellow
MFCD00026494
z-leu-oh, n-cbz-l-leucine, n-carbobenzoxy-l-leucine, carbobenzoxy-l-leucine, n-carbobenzyloxy-l-leucine, carbobenzoxyleucine, z-l-leucine, benzyloxycarbonyl-l-leucine, n-benzyloxycarbonyl-l-leucine, cbz-leu-oh
CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
265.31
CHEBI:28282
≥96.0% (T)
2018-66-8
C14H19NO4
25 g
USPFMEKVPDBMCG-LBPRGKRZSA-N
(2S)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid
74840
265.31
Starch Syrup

Chemical Identifiers

2018-66-8
265.31
USPFMEKVPDBMCG-LBPRGKRZSA-N
74840
(2S)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid
C14H19NO4
MFCD00026494
z-leu-oh, n-cbz-l-leucine, n-carbobenzoxy-l-leucine, carbobenzoxy-l-leucine, n-carbobenzyloxy-l-leucine, carbobenzoxyleucine, z-l-leucine, benzyloxycarbonyl-l-leucine, n-benzyloxycarbonyl-l-leucine, cbz-leu-oh
CHEBI:28282
CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : OH2921000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions