missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-leucine 96.0+%, TCI America™

Catalog No. p-7124586
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07395G 5 g
C073925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07395G Supplier TCI America Supplier No. C07395G
Only null left

Chemical Identifiers

CAS 2018-66-8
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00026494
InChI Key USPFMEKVPDBMCG-LBPRGKRZSA-N
Synonym z-leu-oh, n-cbz-l-leucine, n-carbobenzoxy-l-leucine, carbobenzoxy-l-leucine, n-carbobenzyloxy-l-leucine, carbobenzoxyleucine, z-l-leucine, benzyloxycarbonyl-l-leucine, n-benzyloxycarbonyl-l-leucine, cbz-leu-oh
PubChem CID 74840
ChEBI CHEBI:28282
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 265.31
Percent Purity ≥96.0% (T)
Physical Form Starch Syrup
Chemical Name or Material N-Carbobenzoxy-L-leucine
RTECSNumber OH2921000
TSCA No