accessibility menu, dialog, popup

UserName

H3C(S)HNOHOOOCH3

CAS RN 2018-66-8

IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid
Synonyms: carbobenzoxy-L-leucine|carbobenzyloxy-L-leucine|Z-LEU-OH
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: USPFMEKVPDBMCG-LBPRGKRZSA-N
SMILES: CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results