missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzoxy-3-pyrroline 97.0+%, TCI America™
$100.62 - $482.25
Chemical Identifiers
| CAS | 31970-04-4 |
|---|---|
| Molecular Formula | C12H13NO2 |
| Molecular Weight (g/mol) | 203.241 |
| MDL Number | MFCD01863559 |
| InChI Key | XSKKIFJNZPNVGO-UHFFFAOYSA-N |
| Synonym | benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate, benzyl 3-pyrroline-1-carboxylate, 1-cbz-2,5-dihydro-1h-pyrrole, n-cbz-2,5-dihydropyrrole, n-cbz-3-pyrroline, n-carbobenzoxy-3-pyrroline, 2,5-dihydropyrrole-1-carboxylic acid benzyl ester, benzyl 2h-pyrrole-1 5h-carboxylate, n-benzyloxycarbonyl-3-pyrroline, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester |
| PubChem CID | 643471 |
| IUPAC Name | benzyl 2,5-dihydropyrrole-1-carboxylate |
| SMILES | C1C=CCN1C(=O)OCC2=CC=CC=C2 |
Chemical Identifiers
| 31970-04-4 | |
| 203.241 | |
| XSKKIFJNZPNVGO-UHFFFAOYSA-N | |
| 643471 | |
| C1C=CCN1C(=O)OCC2=CC=CC=C2 |
| C12H13NO2 | |
| MFCD01863559 | |
| benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate, benzyl 3-pyrroline-1-carboxylate, 1-cbz-2,5-dihydro-1h-pyrrole, n-cbz-2,5-dihydropyrrole, n-cbz-3-pyrroline, n-carbobenzoxy-3-pyrroline, 2,5-dihydropyrrole-1-carboxylic acid benzyl ester, benzyl 2h-pyrrole-1 5h-carboxylate, n-benzyloxycarbonyl-3-pyrroline, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester | |
| benzyl 2,5-dihydropyrrole-1-carboxylate |
Specifications
| 31970-04-4 | |
| Yellow | |
| MFCD01863559 | |
| XSKKIFJNZPNVGO-UHFFFAOYSA-N | |
| benzyl 2,5-dihydropyrrole-1-carboxylate | |
| 643471 | |
| ≥97.0% (GC) | |
| N-Carbobenzoxy-3-pyrroline |
| 126°C | |
| C12H13NO2 | |
| benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate, benzyl 3-pyrroline-1-carboxylate, 1-cbz-2,5-dihydro-1h-pyrrole, n-cbz-2,5-dihydropyrrole, n-cbz-3-pyrroline, n-carbobenzoxy-3-pyrroline, 2,5-dihydropyrrole-1-carboxylic acid benzyl ester, benzyl 2h-pyrrole-1 5h-carboxylate, n-benzyloxycarbonyl-3-pyrroline, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester | |
| C1C=CCN1C(=O)OCC2=CC=CC=C2 | |
| 203.241 | |
| 203.24 | |
| Liquid |
Safety and Handling
TSCA : No