missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-3-pyrroline 97.0+%, TCI America™

Catalog No. C28725G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28725G 5 g
C28721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28725G Supplier TCI America Supplier No. C28725G

Chemical Identifiers

CAS 31970-04-4
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD01863559
InChI Key XSKKIFJNZPNVGO-UHFFFAOYSA-N
Synonym benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate, benzyl 3-pyrroline-1-carboxylate, 1-cbz-2,5-dihydro-1h-pyrrole, n-cbz-2,5-dihydropyrrole, n-cbz-3-pyrroline, n-carbobenzoxy-3-pyrroline, 2,5-dihydropyrrole-1-carboxylic acid benzyl ester, benzyl 2h-pyrrole-1 5h-carboxylate, n-benzyloxycarbonyl-3-pyrroline, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester
PubChem CID 643471
IUPAC Name benzyl 2,5-dihydropyrrole-1-carboxylate
SMILES C1C=CCN1C(=O)OCC2=CC=CC=C2

Specifications

Boiling Point 126°C
Color Yellow
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Carbobenzoxy-3-pyrroline