accessibility menu, dialog, popup

UserName

NOO

CAS RN 31970-04-4

IUPAC Name: benzyl 2,5-dihydro-1H-pyrrole-1-carboxylate
Synonyms: benzyl 2,5-dihydropyrrole-1-carboxylate
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: XSKKIFJNZPNVGO-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)N1CC=CC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)