missing translation for 'onlineSavingsMsg'
Learn More

n-Butyllithium, 2.7M solution in toluene, AcroSeal™

Catalog No. p-3073674
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378931000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC378931000 Supplier Thermo Scientific Chemicals Supplier No. 378931000
Only null left

CAS: 109-72-8 | C4H9Li | 64.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-72-8 , 108-88-3
Molecular Formula C4H9Li
Molecular Weight (g/mol) 64.06
MDL Number MFCD00009414
InChI Key MZRVEZGGRBJDDB-UHFFFAOYSA-N
Synonym n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc
PubChem CID 61028
IUPAC Name butyllithium
SMILES [Li]CCCC

Specifications

Color Yellow to Orange
Linear Formula CH3(CH2)3Li
Density 0.8600g/mL
Solubility Information Solubility in water: vigorous reaction.
Formula Weight 64.06
Percent Purity 19.8 to 20.2% (as BuLi)
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.86
Quantity 100 mL
Flash Point 4°C
Physical Form Solution
Chemical Name or Material n-Butyllithium, 2.6M solution in toluene
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Suspected of damaging the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.

RUO – Research Use Only