missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl disulfide, 97%

Catalog No. AC107921000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107921000 100 g Glass bottle
AC107920250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107921000 Supplier Thermo Scientific Chemicals Supplier No. 107921000
Only null left

CAS: 629-45-8 | C8H18S2 | 178.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-45-8
Molecular Formula C8H18S2
Molecular Weight (g/mol) 178.35
MDL Number MFCD00009467
InChI Key CUDSBWGCGSUXDB-UHFFFAOYSA-N
Synonym dibutyl disulfide, butyl disulfide, disulfide, dibutyl, n-butyl disulfide, di-n-butyl disulfide, dibutyl disulphide, 5,6-dithiadecane, 1-butyldisulfanyl butane, unii-z77ef35c0q, 1-butyldisulfanyl-butane
PubChem CID 12386
IUPAC Name 1-(butyldisulfanyl)butane
SMILES CCCCSSCCCC

Specifications

Color Brown to Colorless
Density 0.9380g/mL
Boiling Point 229.0°C to 233.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4915 to 1.4935
Linear Formula [CH3(CH2)3]2S2
Quantity 100 g
Beilstein 01,II,4
Specific Gravity 0.938
Solubility Information Solubility in water: immiscible
Formula Weight 178.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material n-Butyl disulfide
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 211-091-3

RUO – Research Use Only