missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl benzoate, 98%

CAS: 136-60-7 | C11H14O2 | 178.23 g/mol

Supplier:  Thermo Scientific Chemicals A170100B

 View more versions of this product

Catalog No. AAA170100B


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Butyl benzoate
-22°C
250°C
C11H14O2
MFCD00009439
1867073
n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech
CCCCOC(=O)C1=CC=CC=C1
178.23
178.23
136-60-7
1.011
106°C (222°F)
1.498
1000 g
14,1552
XSIFPSYPOVKYCO-UHFFFAOYSA-N
butyl benzoate
8698
98%

Chemical Identifiers

136-60-7
178.23
XSIFPSYPOVKYCO-UHFFFAOYSA-N
8698
CCCCOC(=O)C1=CC=CC=C1
C11H14O2
MFCD00009439
n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech
butyl benzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 205-252-7

RTECSNumber : DG4925000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only