missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl benzoate, 98%

Catalog No. p-7023447
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1701030 250 g
AAA170100B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1701030 Supplier Thermo Scientific Chemicals Supplier No. A1701030
Only null left

CAS: 136-60-7 | C11H14O2 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136-60-7
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00009439
InChI Key XSIFPSYPOVKYCO-UHFFFAOYSA-N
Synonym n-butyl benzoate, benzoic acid, butyl ester, anthrapole az, benzoic acid butyl ester, dai cari xbn, benzoic acid n-butyl ester, butylester kyseliny benzoove, butylbenzoate, unii-1tgz0d0o8i, butylester kyseliny benzoove czech
PubChem CID 8698
IUPAC Name butyl benzoate
SMILES CCCCOC(=O)C1=CC=CC=C1

Specifications

Melting Point -22°C
Density 1.011
Boiling Point 250°C
Flash Point 106°C (222°F)
Refractive Index 1.498
Quantity 250 g
Beilstein 1867073
Merck Index 14,1552
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material n-Butyl benzoate
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 205-252-7
RTECSNumber DG4925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only