missing translation for 'onlineSavingsMsg'
Learn More

n-Butanol, 7.5% v/v, Fisher Chemical™

Catalog No. SB95754
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB95754 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB95754 Supplier Ricca Chemical Company Supplier No. SB95754

Chemical Identifiers

CAS 71-36-3 , 7732-18-5 , 497-19-8 , 144-55-8
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

CAS Min % 92.0, 5.54, 8.0E-4, 7.0E-4
CAS Max % 96.0, 5.77, 8.0E-4, 7.0E-4
Quantity 4 L
Packaging Amber Glass Bottle
Chemical Name or Material n-Butanol, 7.5% v/v