Learn More
N-Bromoacetamide, 95%
CAS: 79-15-2 | C2H4BrNO | 137.964 g/mol
Supplier: Thermo Scientific Chemicals L0295314
Description
Abolishes the rapid inactivation of membrane sodium- and potassium-ion channels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAbolishes the rapid inactivation of membrane sodium- and potassium-ion channels.
Solubility
Soluble in water.
Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| N-Bromoacetamide | |
| 102°C to 106°C | |
| CH3CONHBr | |
| 25 g | |
| 14,1398 | |
| Soluble in water. | |
| CC(=O)NBr | |
| 137.964 | |
| 137.97 |
| 79-15-2 | |
| C2H4BrNO | |
| MFCD00037097 | |
| 969346 | |
| acetamide, n-bromo, unii-0aq6mwh7zb, ccris 4590, 0aq6mwh7zb, acetobromamide, n-bromo-acetamide, sjyhiabiktp@, acmc-209pgg, n-bromoacetamide, powder, lopac-b-2377 | |
| VBTQNRFWXBXZQR-UHFFFAOYSA-N | |
| N-bromoacetamide | |
| 4353 | |
| 95% |
Chemical Identifiers
| 79-15-2 | |
| 137.964 | |
| VBTQNRFWXBXZQR-UHFFFAOYSA-N | |
| 4353 | |
| CC(=O)NBr |
| C2H4BrNO | |
| MFCD00037097 | |
| acetamide, n-bromo, unii-0aq6mwh7zb, ccris 4590, 0aq6mwh7zb, acetobromamide, n-bromo-acetamide, sjyhiabiktp@, acmc-209pgg, n-bromoacetamide, powder, lopac-b-2377 | |
| N-bromoacetamide |
Safety and Handling
GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 201-181-0
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only