missing translation for 'onlineSavingsMsg'
Learn More

N-Bromoacetamide, 95%

Catalog No. p-7043830
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0295314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0295314 Supplier Thermo Scientific Chemicals Supplier No. L0295314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-15-2 | C2H4BrNO | 137.964 g/mol

Abolishes the rapid inactivation of membrane sodium- and potassium-ion channels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Abolishes the rapid inactivation of membrane sodium- and potassium-ion channels.

Solubility
Soluble in water.

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 79-15-2
Molecular Formula C2H4BrNO
Molecular Weight (g/mol) 137.964
MDL Number MFCD00037097
InChI Key VBTQNRFWXBXZQR-UHFFFAOYSA-N
Synonym acetamide, n-bromo, unii-0aq6mwh7zb, ccris 4590, 0aq6mwh7zb, acetobromamide, n-bromo-acetamide, sjyhiabiktp@, acmc-209pgg, n-bromoacetamide, powder, lopac-b-2377
PubChem CID 4353
IUPAC Name N-bromoacetamide
SMILES CC(=O)NBr

Specifications

Melting Point 102°C to 106°C
Linear Formula CH3CONHBr
Quantity 25 g
Beilstein 969346
Merck Index 14,1398
Solubility Information Soluble in water.
Formula Weight 137.97
Percent Purity 95%
Chemical Name or Material N-Bromoacetamide
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 201-181-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only