missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-S-trityl-L-cysteine, 97%

Catalog No. AAH5949006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5949006 5 g
AAH5949014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5949006 Supplier Thermo Scientific Chemicals Supplier No. H5949006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 21947-98-8 | C27H29NO4S | 463.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21947-98-8
Molecular Formula C27H29NO4S
Molecular Weight (g/mol) 463.592
MDL Number MFCD00038251
InChI Key JDTOWOURWBDELG-QHCPKHFHSA-N
Synonym boc-cys trt-oh, n-boc-s-trityl-l-cysteine, boc-s-trityl-l-cysteine, n-tert-butoxycarbonyl-s-trityl-l-cysteine, r-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-cysteine trt-oh, 2r-2-tert-butoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-triphenylmethyl, boc-l-cysteine trityl, 2r-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid
PubChem CID 11167161
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-tritylsulfanylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O

Specifications

Melting Point 143°C to 146°C
Quantity 5 g
Beilstein 2028965
Optical Rotation +28° (c=1 in Ethanol)
Formula Weight 463.59
Percent Purity 97%
Chemical Name or Material N-Boc-S-trityl-L-cysteine
EINECSNumber 244-674-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only