accessibility menu, dialog, popup

UserName

CAS RN 21947-98-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-[(triphenylmethyl)sulfanyl]propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-[(triphenylmethyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 463.59
Molecular Formula: C27H29NO4S
InChi Key: JDTOWOURWBDELG-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results