missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-glutamic acid 5-methyl ester, 95%
CAS: 45214-91-3 | C11H19NO6 | 261.274 g/mol
Supplier: Thermo Scientific Chemicals H6249606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Boc-L-glutamic acid 5-methyl ester | |
| White | |
| MFCD00190794 | |
| OHYMUFVCRVPMEY-ZETCQYMHSA-N | |
| (2S)-5-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid | |
| 261.274 | |
| 261.27 | |
| Solid |
| 45214-91-3 | |
| C11H19NO6 | |
| boc-glu ome-oh, s-2-tert-butoxycarbonyl amino-5-methoxy-5-oxopentanoic acid, boc-l-glutamic acid gamma-methyl ester, s-2-tert-butoxycarbonylamino-5-methoxy-5-oxopentanoic acid, 2s-2-tert-butoxycarbonyl amino-5-methoxy-5-oxopentanoic acid, pubchem18618, boc-l-glutamic acid methyl ester, n-boc-l-glutamic acid 5-methyl ester, n-t-butoxycarbonyl-l-glutamic acid 5-methyl ester, n-tert-butoxycarbonylglutamic acid ?-methyl ester | |
| CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)O | |
| 5 g | |
| 7018756 | |
| 95% |
Chemical Identifiers
| 45214-91-3 | |
| 261.274 | |
| OHYMUFVCRVPMEY-ZETCQYMHSA-N | |
| 7018756 | |
| CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)O |
| C11H19NO6 | |
| MFCD00190794 | |
| boc-glu ome-oh, s-2-tert-butoxycarbonyl amino-5-methoxy-5-oxopentanoic acid, boc-l-glutamic acid gamma-methyl ester, s-2-tert-butoxycarbonylamino-5-methoxy-5-oxopentanoic acid, 2s-2-tert-butoxycarbonyl amino-5-methoxy-5-oxopentanoic acid, pubchem18618, boc-l-glutamic acid methyl ester, n-boc-l-glutamic acid 5-methyl ester, n-t-butoxycarbonyl-l-glutamic acid 5-methyl ester, n-tert-butoxycarbonylglutamic acid ?-methyl ester | |
| (2S)-5-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only