missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-glutamic acid 5-methyl ester, 95%

Catalog No. p-7054357
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62496MD 250 mg
AAH6249603 1 g
AAH6249606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62496MD Supplier Thermo Scientific Chemicals Supplier No. H62496MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 45214-91-3 | C11H19NO6 | 261.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 45214-91-3
Molecular Formula C11H19NO6
Molecular Weight (g/mol) 261.274
MDL Number MFCD00190794
InChI Key OHYMUFVCRVPMEY-ZETCQYMHSA-N
Synonym boc-glu ome-oh, s-2-tert-butoxycarbonyl amino-5-methoxy-5-oxopentanoic acid, boc-l-glutamic acid gamma-methyl ester, s-2-tert-butoxycarbonylamino-5-methoxy-5-oxopentanoic acid, 2s-2-tert-butoxycarbonyl amino-5-methoxy-5-oxopentanoic acid, pubchem18618, boc-l-glutamic acid methyl ester, n-boc-l-glutamic acid 5-methyl ester, n-t-butoxycarbonyl-l-glutamic acid 5-methyl ester, n-tert-butoxycarbonylglutamic acid ?-methyl ester
PubChem CID 7018756
IUPAC Name (2S)-5-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)O

Specifications

Color White
Quantity 250 mg
Formula Weight 261.27
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-L-glutamic acid 5-methyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only