missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-aspartic acid 4-tert-butyl ester, 98%
CAS: 1676-90-0 | C13H23NO6 | 289.328 g/mol
$98.53 - $363.86
Chemical Identifiers
| CAS | 1676-90-0 |
|---|---|
| Molecular Formula | C13H23NO6 |
| Molecular Weight (g/mol) | 289.328 |
| MDL Number | MFCD00076912 |
| InChI Key | PHJDCONJXLIIPW-QMMMGPOBSA-N |
| Synonym | boc-asp otbu-oh, boc-l-aspartic acid 4-tert-butyl ester, boc-asp obut-oh, s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-l-aspartic acid beta-t-butyl ester, 2s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp but, pubchem12101, n-boc-asp o-t-bu oh |
| PubChem CID | 7010636 |
| IUPAC Name | (2S)-4-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
| SMILES | CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6355406
|
Thermo Scientific Chemicals
H6355406 |
5 g |
Each for $98.53
|
|
|||||
|
AAH6355414
|
Thermo Scientific Chemicals
H6355414 |
25 g |
Each for $363.86
|
|
|||||
Description
It is used in the preparation of highly selective thrombin inhibitors. Also used in the preparation of thiopeptides and thioacylating agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the preparation of highly selective thrombin inhibitors. Also used in the preparation of thiopeptides and thioacylating agents.
Solubility
Soluble in methanol and dimethyl sulfoxide.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 1676-90-0 | |
| 289.328 | |
| PHJDCONJXLIIPW-QMMMGPOBSA-N | |
| 7010636 | |
| CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OC(C)(C)C |
| C13H23NO6 | |
| MFCD00076912 | |
| boc-asp otbu-oh, boc-l-aspartic acid 4-tert-butyl ester, boc-asp obut-oh, s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-l-aspartic acid beta-t-butyl ester, 2s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp but, pubchem12101, n-boc-asp o-t-bu oh | |
| (2S)-4-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
Specifications
| 64°C to 67°C | |
| White | |
| MFCD00076912 | |
| boc-asp otbu-oh, boc-l-aspartic acid 4-tert-butyl ester, boc-asp obut-oh, s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-l-aspartic acid beta-t-butyl ester, 2s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp but, pubchem12101, n-boc-asp o-t-bu oh | |
| PHJDCONJXLIIPW-QMMMGPOBSA-N | |
| (2S)-4-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid | |
| 289.328 | |
| 289.32 | |
| Powder |
| 1676-90-0 | |
| C13H23NO6 | |
| 2336144 | |
| Soluble in methanol and dimethyl sulfoxide. | |
| CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OC(C)(C)C | |
| 5 g | |
| 7010636 | |
| 98% | |
| N-Boc-L-aspartic acid 4-tert-butyl ester |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only