missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-aspartic acid 4-tert-butyl ester, 98%

CAS: 1676-90-0 | C13H23NO6 | 289.328 g/mol

Supplier:  Thermo Scientific Chemicals H6355414

 View more versions of this product

Catalog No. AAH6355414


Only null left

Description

Description

It is used in the preparation of highly selective thrombin inhibitors. Also used in the preparation of thiopeptides and thioacylating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of highly selective thrombin inhibitors. Also used in the preparation of thiopeptides and thioacylating agents.

Solubility
Soluble in methanol and dimethyl sulfoxide.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

64°C to 67°C
1676-90-0
C13H23NO6
2336144
Soluble in methanol and dimethyl sulfoxide.
CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OC(C)(C)C
25 g
7010636
98%
N-Boc-L-aspartic acid 4-tert-butyl ester
White
MFCD00076912
boc-asp otbu-oh, boc-l-aspartic acid 4-tert-butyl ester, boc-asp obut-oh, s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-l-aspartic acid beta-t-butyl ester, 2s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp but, pubchem12101, n-boc-asp o-t-bu oh
PHJDCONJXLIIPW-QMMMGPOBSA-N
(2S)-4-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
289.328
289.32
Powder

Chemical Identifiers

1676-90-0
289.328
PHJDCONJXLIIPW-QMMMGPOBSA-N
7010636
CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OC(C)(C)C
C13H23NO6
MFCD00076912
boc-asp otbu-oh, boc-l-aspartic acid 4-tert-butyl ester, boc-asp obut-oh, s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-l-aspartic acid beta-t-butyl ester, 2s-4-tert-butoxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp but, pubchem12101, n-boc-asp o-t-bu oh
(2S)-4-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only