accessibility menu, dialog, popup

UserName

H3CCH3H3COOOHONHOOCH3CH3H3C

CAS RN 1676-90-0

IUPAC Name: 4-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}-4-oxobutanoic acid
Synonyms: 4-(tert-butoxy)-2-[(tert-butoxycarbonyl)amino]-4-oxobutanoic acid
Molecular Weight (g/mol): 289.33
Molecular Formula: C13H23NO6
InChi Key: PHJDCONJXLIIPW-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CC(NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)