missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-glycine ethyl ester, 95%

CAS: 14719-37-0 | C9H17NO4 | 203.238 g/mol

Supplier:  Thermo Scientific Chemicals H6261106

 View more versions of this product

Catalog No. AAH6261106


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Boc-glycine ethyl ester
Colorless
(CH3)3COCONHCH2CO2CH2CH3
n-boc-glycine ethyl ester, ethyl 2-tert-butoxycarbonyl amino acetate, ethyl 2-tert-butoxycarbonylamino acetate, boc-glycine ethyl ester, boc-gly-oet, tert-butoxycarbonylamino-acetic acid ethyl ester, ethyl boc-aminoacetate, n-tert-butoxycarbonyl glycine ethyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, ethyl ester, ethyl 2-tert-butoxy carbonyl amino acetate
CCOC(=O)CNC(=O)OC(C)(C)C
5 g
357729
95%
14719-37-0
C9H17NO4
MFCD05663720
CNIBHMMDDXGDNR-UHFFFAOYSA-N
ethyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
203.238
203.24
Oily Liquid

Chemical Identifiers

14719-37-0
203.238
CNIBHMMDDXGDNR-UHFFFAOYSA-N
357729
CCOC(=O)CNC(=O)OC(C)(C)C
C9H17NO4
MFCD05663720
n-boc-glycine ethyl ester, ethyl 2-tert-butoxycarbonyl amino acetate, ethyl 2-tert-butoxycarbonylamino acetate, boc-glycine ethyl ester, boc-gly-oet, tert-butoxycarbonylamino-acetic acid ethyl ester, ethyl boc-aminoacetate, n-tert-butoxycarbonyl glycine ethyl ester, glycine, n-1,1-dimethylethoxy carbonyl-, ethyl ester, ethyl 2-tert-butoxy carbonyl amino acetate
ethyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only