accessibility menu, dialog, popup

UserName

CAS RN 14719-37-0

IUPAC Name: ethyl 2-{[(tert-butoxy)carbonyl]amino}acetate
Synonyms: ethyl 2-[(tert-butoxycarbonyl)amino]acetate
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: CNIBHMMDDXGDNR-UHFFFAOYSA-N
SMILES: CCOC(=O)CNC(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)