missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-serine, 98+%

CAS: 6368-20-3 | C8H15NO5 | 205.21 g/mol

Supplier:  Thermo Scientific Chemicals L0890406

 View more versions of this product

Catalog No. AAL0890406


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Boc-D-serine
88°C to 91°C
MFCD00063142
1874714
FHOAKXBXYSJBGX-RXMQYKEDSA-N
CC(C)(C)OC(=O)NC(CO)C(=O)O
205.21
205.21
6368-20-3
C8H15NO5
5 g
boc-d-serine, boc-d-ser-oh, n-boc-d-serine, n-tert-butoxycarbonyl-d-serine, n-alpha-t-butyloxycarbonyl-d-serine, boc-l-serine, boc-l-ser-oh, 2r-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, d-serine, n-1,1-dimethylethoxy carbonyl, r-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid
+3.5° (c=2 in Acetic Acid)
(2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
637603
≥98%

Chemical Identifiers

6368-20-3
205.21
FHOAKXBXYSJBGX-RXMQYKEDSA-N
637603
CC(C)(C)OC(=O)NC(CO)C(=O)O
C8H15NO5
MFCD00063142
boc-d-serine, boc-d-ser-oh, n-boc-d-serine, n-tert-butoxycarbonyl-d-serine, n-alpha-t-butyloxycarbonyl-d-serine, boc-l-serine, boc-l-ser-oh, 2r-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, d-serine, n-1,1-dimethylethoxy carbonyl, r-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid
(2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 613-313-8

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only