missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-serine, 98+%

Catalog No. p-7044966
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0890403 1 g
AAL0890406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0890403 Supplier Thermo Scientific Chemicals Supplier No. L0890403
Only null left

CAS: 6368-20-3 | C8H15NO5 | 205.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6368-20-3
Molecular Formula C8H15NO5
Molecular Weight (g/mol) 205.21
MDL Number MFCD00063142
InChI Key FHOAKXBXYSJBGX-RXMQYKEDSA-N
Synonym boc-d-serine, boc-d-ser-oh, n-boc-d-serine, n-tert-butoxycarbonyl-d-serine, n-alpha-t-butyloxycarbonyl-d-serine, boc-l-serine, boc-l-ser-oh, 2r-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, d-serine, n-1,1-dimethylethoxy carbonyl, r-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid
PubChem CID 637603
IUPAC Name (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)O

Specifications

Melting Point 88°C to 91°C
Quantity 1 g
Beilstein 1874714
Optical Rotation +3.5° (c=2 in Acetic Acid)
Formula Weight 205.21
Percent Purity ≥98%
Chemical Name or Material N-Boc-D-serine
EINECSNumber 613-313-8
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only