missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-D-aspartic acid 1-benzyl ester, 98%
CAS: 92828-64-3 | C16H21NO6 | 323.35 g/mol
$136.84 - $530.99
Chemical Identifiers
| CAS | 92828-64-3 |
|---|---|
| Molecular Formula | C16H21NO6 |
| Molecular Weight (g/mol) | 323.35 |
| MDL Number | MFCD00038264 |
| InChI Key | LDRWTKQWSXGSTM-UHFFFAOYNA-N |
| Synonym | boc-d-asp-obzl, r-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, 3r-4-benzyloxy-3-tert-butoxy carbonyl amino-4-oxobutanoic acid, 3r-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, d-aspartic acid,n-1,1-dimethylethoxy carbonyl-, 1-phenylmethyl ester, boc-d-aspartic acid a-benzyl ester, boc-d-aspartic acid-1-benzyl ester, boc-d-aspartic acid, a-benzyl ester, boc-d-aspartic acid benzylester, n-tert-butoxycarbonyl-d-aspartic acid 1-benzyl ester |
| PubChem CID | 11759109 |
| SMILES | CC(C)(C)OC(=O)NC(CC(O)=O)C(=O)OCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH6221003
|
N/A | 1 g |
Each for $136.84
|
|
|||||
|
AAH6221006
|
N/A | 5 g |
Each for $530.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 92828-64-3 | |
| 323.35 | |
| LDRWTKQWSXGSTM-UHFFFAOYNA-N | |
| 11759109 |
| C16H21NO6 | |
| MFCD00038264 | |
| boc-d-asp-obzl, r-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, 3r-4-benzyloxy-3-tert-butoxy carbonyl amino-4-oxobutanoic acid, 3r-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, d-aspartic acid,n-1,1-dimethylethoxy carbonyl-, 1-phenylmethyl ester, boc-d-aspartic acid a-benzyl ester, boc-d-aspartic acid-1-benzyl ester, boc-d-aspartic acid, a-benzyl ester, boc-d-aspartic acid benzylester, n-tert-butoxycarbonyl-d-aspartic acid 1-benzyl ester | |
| CC(C)(C)OC(=O)NC(CC(O)=O)C(=O)OCC1=CC=CC=C1 |
Specifications
| 92828-64-3 | |
| C16H21NO6 | |
| boc-d-asp-obzl, r-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, 3r-4-benzyloxy-3-tert-butoxy carbonyl amino-4-oxobutanoic acid, 3r-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, d-aspartic acid,n-1,1-dimethylethoxy carbonyl-, 1-phenylmethyl ester, boc-d-aspartic acid a-benzyl ester, boc-d-aspartic acid-1-benzyl ester, boc-d-aspartic acid, a-benzyl ester, boc-d-aspartic acid benzylester, n-tert-butoxycarbonyl-d-aspartic acid 1-benzyl ester | |
| CC(C)(C)OC(=O)NC(CC(O)=O)C(=O)OCC1=CC=CC=C1 | |
| 323.35 | |
| 323.34 | |
| Powder |
| White | |
| MFCD00038264 | |
| LDRWTKQWSXGSTM-UHFFFAOYNA-N | |
| (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid | |
| 11759109 | |
| 98% | |
| N-Boc-D-aspartic acid 1-benzyl ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only