missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-aspartic acid 1-benzyl ester, 98%

Catalog No. AAH6221003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6221003 1 g
AAH6221006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6221003 Supplier Thermo Scientific Chemicals Supplier No. H6221003
Only null left

CAS: 92828-64-3 | C16H21NO6 | 323.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92828-64-3
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.35
MDL Number MFCD00038264
InChI Key LDRWTKQWSXGSTM-UHFFFAOYNA-N
Synonym boc-d-asp-obzl, r-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, 3r-4-benzyloxy-3-tert-butoxy carbonyl amino-4-oxobutanoic acid, 3r-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, d-aspartic acid,n-1,1-dimethylethoxy carbonyl-, 1-phenylmethyl ester, boc-d-aspartic acid a-benzyl ester, boc-d-aspartic acid-1-benzyl ester, boc-d-aspartic acid, a-benzyl ester, boc-d-aspartic acid benzylester, n-tert-butoxycarbonyl-d-aspartic acid 1-benzyl ester
PubChem CID 11759109
SMILES CC(C)(C)OC(=O)NC(CC(O)=O)C(=O)OCC1=CC=CC=C1

Specifications

Color White
IUPAC Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
Quantity 1 g
Formula Weight 323.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Boc-D-aspartic acid 1-benzyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only