missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-alaninol, 98%, ee 98%

CAS: 106391-86-0 | C8H17NO3 | 175.23 g/mol

$66.32 - $663.30

Chemical Identifiers

CAS 106391-86-0
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD00235912
InChI Key PDAFIZPRSXHMCO-ZCFIWIBFSA-N
Synonym n-boc-d-alaninol, boc-d-alaninol, r-2-boc-amino-1-propanol, tert-butyl n-2r-1-hydroxypropan-2-yl carbamate, r-tert-butyl 1-hydroxypropan-2-yl carbamate, n-alpha-t-boc-d-alaninol, r-tert-butyl 1-hydroxypropan-2-ylcarbamate, r-+-2-tert-butoxycarbonylamino-1-propanol, tert-butyl 2r-1-hydroxypropan-2-yl carbamate, tert-butyl n-1r-2-hydroxy-1-methyl-ethyl carbamate
PubChem CID 13200309
IUPAC Name tert-butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
SMILES C[C@H](CO)NC(=O)OC(C)(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH2788703
View Documents Promotion Details
N/A 1 g
Each for $66.32
Only null left
 
AAH2788706
View Documents Promotion Details
N/A 5 g
Each for $210.66
Only null left
 
AAH2788714
View Documents Promotion Details
N/A 25 g
Each for $663.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106391-86-0
175.23
PDAFIZPRSXHMCO-ZCFIWIBFSA-N
13200309
C[C@H](CO)NC(=O)OC(C)(C)C
C8H17NO3
MFCD00235912
n-boc-d-alaninol, boc-d-alaninol, r-2-boc-amino-1-propanol, tert-butyl n-2r-1-hydroxypropan-2-yl carbamate, r-tert-butyl 1-hydroxypropan-2-yl carbamate, n-alpha-t-boc-d-alaninol, r-tert-butyl 1-hydroxypropan-2-ylcarbamate, r-+-2-tert-butoxycarbonylamino-1-propanol, tert-butyl 2r-1-hydroxypropan-2-yl carbamate, tert-butyl n-1r-2-hydroxy-1-methyl-ethyl carbamate
tert-butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate

Specifications

58°C to 62°C
C8H17NO3
n-boc-d-alaninol, boc-d-alaninol, r-2-boc-amino-1-propanol, tert-butyl n-2r-1-hydroxypropan-2-yl carbamate, r-tert-butyl 1-hydroxypropan-2-yl carbamate, n-alpha-t-boc-d-alaninol, r-tert-butyl 1-hydroxypropan-2-ylcarbamate, r-+-2-tert-butoxycarbonylamino-1-propanol, tert-butyl 2r-1-hydroxypropan-2-yl carbamate, tert-butyl n-1r-2-hydroxy-1-methyl-ethyl carbamate
C[C@H](CO)NC(=O)OC(C)(C)C
175.23
+11° (c=1 in Chloroform)
98%
N-Boc-D-alaninol
106391-86-0
MFCD00235912
PDAFIZPRSXHMCO-ZCFIWIBFSA-N
tert-butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
13200309
175.23
ee 98%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only