missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-alaninol, 98%, ee 98%

Catalog No. AAH2788714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2788714 25 g
AAH2788703 1 g
AAH2788706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2788714 Supplier Thermo Scientific Chemicals Supplier No. H2788714

CAS: 106391-86-0 | C8H17NO3 | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106391-86-0
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD00235912
InChI Key PDAFIZPRSXHMCO-ZCFIWIBFSA-N
Synonym n-boc-d-alaninol, boc-d-alaninol, r-2-boc-amino-1-propanol, tert-butyl n-2r-1-hydroxypropan-2-yl carbamate, r-tert-butyl 1-hydroxypropan-2-yl carbamate, n-alpha-t-boc-d-alaninol, r-tert-butyl 1-hydroxypropan-2-ylcarbamate, r-+-2-tert-butoxycarbonylamino-1-propanol, tert-butyl 2r-1-hydroxypropan-2-yl carbamate, tert-butyl n-1r-2-hydroxy-1-methyl-ethyl carbamate
PubChem CID 13200309
IUPAC Name tert-butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
SMILES C[C@H](CO)NC(=O)OC(C)(C)C

Specifications

Melting Point 58°C to 62°C
Quantity 25 g
Optical Rotation +11° (c=1 in Chloroform)
Formula Weight 175.23
Percent Purity 98%
Assay ee 98%
Chemical Name or Material N-Boc-D-alaninol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only