CAS RN 106391-86-0
IUPAC Name:
tert-butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
Synonyms:
tert-butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate|
N-BOC-D-ALANINOL
Molecular Weight (g/mol):
175.23
Molecular Formula:
C8H17NO3
InChi Key:
PDAFIZPRSXHMCO-ZCFIWIBFSA-N
SMILES:
C[C@H](CO)NC(=O)OC(C)(C)C
1
–
1
of
1
results
1
N-Boc-D-alaninol, 98%, ee 98%
CAS: 106391-86-0 Molecular Formula: C8H17NO3 Molecular Weight (g/mol): 175.23 MDL Number: MFCD00235912 InChI Key: PDAFIZPRSXHMCO-ZCFIWIBFSA-N Synonym: n-boc-d-alaninol,boc-d-alaninol,r-2-boc-amino-1-propanol,tert-butyl n-2r-1-hydroxypropan-2-yl carbamate,r-tert-butyl 1-hydroxypropan-2-yl carbamate,n-alpha-t-boc-d-alaninol,r-tert-butyl 1-hydroxypropan-2-ylcarbamate,r-+-2-tert-butoxycarbonylamino-1-propanol,tert-butyl 2r-1-hydroxypropan-2-yl carbamate,tert-butyl n-1r-2-hydroxy-1-methyl-ethyl carbamate PubChem CID: 13200309 IUPAC Name: tert-butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate SMILES: C[C@H](CO)NC(=O)OC(C)(C)C