Learn More
N-Boc-1,6-diaminohexane, 98%, Thermo Scientific Chemicals
$88.90 - $969.92
Chemical Identifiers
CAS | 51857-17-1 |
---|---|
Molecular Formula | C11H24N2O2 |
Molecular Weight (g/mol) | 216.325 |
MDL Number | MFCD00671489 |
InChI Key | RVZPDKXEHIRFPM-UHFFFAOYSA-N |
Synonym | n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl ester |
PubChem CID | 2733170 |
IUPAC Name | tert-butyl N-(6-aminohexyl)carbamate |
SMILES | CC(C)(C)OC(=O)NCCCCCCN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1994803
|
Thermo Scientific Chemicals
L1994803 |
1 g |
Each for $88.90
|
|
|||||
AAL1994806
|
Thermo Scientific Chemicals
L1994806 |
5 g |
Each for $276.16
|
|
|||||
AAL1994814
|
Thermo Scientific Chemicals
L1994814 |
25 g |
Each for $969.92
|
|
|||||
Description
N-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.
Solubility
Soluble in dichloromethane and ethyl acetate.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
51857-17-1 | |
216.325 | |
RVZPDKXEHIRFPM-UHFFFAOYSA-N | |
2733170 | |
CC(C)(C)OC(=O)NCCCCCCN |
C11H24N2O2 | |
MFCD00671489 | |
n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl ester | |
tert-butyl N-(6-aminohexyl)carbamate |
Specifications
51857-17-1 | |
C11H24N2O2 | |
UN2735 | |
n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl ester | |
Soluble in dichloromethane and ethyl acetate. | |
CC(C)(C)OC(=O)NCCCCCCN | |
216.325 | |
216.33 | |
0.965 | |
>110°C (230°F) | |
N-Boc-1,6-diaminohexane |
106°C to 110°C (0.3mmHg) | |
MFCD00671489 | |
2089264 | |
1.461 | |
RVZPDKXEHIRFPM-UHFFFAOYSA-N | |
tert-butyl N-(6-aminohexyl)carbamate | |
2733170 | |
98% | |
1 g | |
Air Sensitive |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 628-984-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only