missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,6-diaminohexane, 98%

CAS: 51857-17-1 | C11H24N2O2 | 216.325 g/mol

Supplier:  Thermo Scientific Chemicals L1994806

 View more versions of this product

Catalog No. AAL1994806


Only null left

Description

Description

N-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.

Solubility
Soluble in dichloromethane and ethyl acetate.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

N-Boc-1,6-diaminohexane
106°C to 110°C (0.3mmHg)
1.461
UN2735
n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl ester
RVZPDKXEHIRFPM-UHFFFAOYSA-N
CC(C)(C)OC(=O)NCCCCCCN
216.325
>110°C (230°F)
216.33
Air Sensitive
51857-17-1
C11H24N2O2
MFCD00671489
2089264
Soluble in dichloromethane and ethyl acetate.
0.965
tert-butyl N-(6-aminohexyl)carbamate
5 g
2733170
98%

Chemical Identifiers

51857-17-1
216.325
RVZPDKXEHIRFPM-UHFFFAOYSA-N
2733170
CC(C)(C)OC(=O)NCCCCCCN
C11H24N2O2
MFCD00671489
n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl ester
tert-butyl N-(6-aminohexyl)carbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 628-984-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only